Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135703.2 | NP_001129175.1 | 4 | 921 | ||
NM_013437.4 | NP_038465.1 | 5 | 978 | |||
GenBank mRNA | AB208957.1 | - | 3 | 653 | ||
AF166350.1 | - | 5 | 893 | |||
AK289752.1 | - | 5 | 882 | |||
AK299374.1 | - | 4 | 918 | |||
BC032109.2 | - | 5 | 854 | 4842797 |