Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197107.1 | NP_001184036.1 | 4 | 1427 | ||
NM_001197108.1 | NP_001184037.1 | 3 | 770 | |||
NM_001197109.1 | NP_001184038.1 | 3 | 487 | |||
NM_014575.3 | NP_055390.1 | 4 | 1466 | |||
GenBank mRNA | AF070614.1 | - | 3 | 576 | ||
AF145713.1 | - | 4 | 961 | |||
AF145714.1 | - | 3 | 265 | |||
AF145715.1 | - | 4 | 922 | |||
AK091871.1 | - | 3 | 408 | |||
AK309223.1 | - | 4 | 874 | |||
AK314662.1 | - | 4 | 1409 | |||
BC005947.1 | - | 3 | 303 | 4246326 | ||
BC036535.2 | - | 4 | 1466 | 5263291 | ||
BX648179.1 | - | 3 | 601 |
Gene Symbol: IQCJ-SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197113.1 | NP_001184042.1 | 7 | 1301 | ||
NM_001197114.1 | NP_001184043.1 | 6 | 1220 | |||
GenBank mRNA | AK096479.1 | - | 7 | 1207 | ||
DQ157847.1 | - | 5 | 1039 | |||
DQ157848.1 | - | 6 | 1120 |