Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030759.4 | NP_110386.2 | 2 | 276 | ||
XM_006717810.3 | XP_006717873.1 | 2 | 238 | |||
GenBank mRNA | AF267866.1 | - | 2 | 227 | ||
AK054957.1 | - | 2 | 234 | |||
AK225256.1 | - | 2 | 185 | |||
AK225257.1 | - | 2 | 187 | |||
AL136656.1 | - | 2 | 231 | |||
AU310836.1 | - | 2 | 276 | |||
AY267839.1 | - | 2 | 164 | |||
AY267840.1 | - | 2 | 164 | |||
BC011707.1 | - | 2 | 150 | 3687848 |