Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282405.1 | NP_001269334.1 | 3 | 432 | ||
NM_030759.4 | NP_110386.2 | 4 | 517 | |||
XM_006717810.3 | XP_006717873.1 | 5 | 702 | |||
GenBank mRNA | AF267866.1 | - | 4 | 468 | ||
AK054957.1 | - | 4 | 475 | |||
AK225256.1 | - | 4 | 426 | |||
AK225257.1 | - | 4 | 428 | |||
AK301656.1 | - | 3 | 354 | |||
AL136656.1 | - | 4 | 472 | |||
AU310836.1 | - | 4 | 517 | |||
AY267840.1 | - | 4 | 405 | |||
BC001345.1 | - | 1 | 101 | |||
BC011707.1 | - | 4 | 391 | 3687848 |