Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014578.3 | NP_055393.1 | 3 | 388 | ||
GenBank mRNA | AF498973.1 | - | 3 | 303 | ||
BC001338.1 | - | 3 | 356 | 3461774 | ||
BM761353.1 | - | 3 | 388 | |||
BM761362.1 | - | 3 | 388 | |||
LN607835.1 | - | 1 | 82 |