Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034837.2 | NP_001030009.1 | 7 | 1056 | ||
NM_001034838.2 | NP_001030010.1 | 6 | 903 | |||
NM_001278339.1 | NP_001265268.1 | 6 | 1098 | |||
NM_001278340.1 | NP_001265269.1 | 7 | 1185 | |||
NM_014592.3 | NP_055407.1 | 6 | 1023 | |||
XM_017009407.1 | XP_016864896.1 | 7 | 1027 | |||
GenBank mRNA | AF199597.1 | - | 6 | 486 | ||
AK074390.1 | - | 6 | 619 | |||
AK301775.1 | - | 6 | 678 | |||
AY170821.1 | - | 6 | 699 | |||
AY780424.1 | - | 7 | 861 | |||
BC035032.1 | - | 6 | 1028 | |||
BC050375.1 | - | 6 | 790 | 5752207 | ||
DQ148476.1 | - | 6 | 513 | |||
DQ148477.1 | - | 6 | 486 | |||
DQ148478.1 | - | 7 | 519 | |||
DQ148479.1 | - | 6 | 561 |