Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 822 | ||
NM_001144894.1 | NP_001138366.1 | 5 | 1098 | |||
NM_001144895.1 | NP_001138367.1 | 7 | 954 | |||
NM_001144896.1 | NP_001138368.1 | 6 | 1158 | |||
NM_001144897.1 | NP_001138369.1 | 7 | 1212 | |||
NM_001144899.1 | NP_001138371.1 | 7 | 747 | |||
NM_021155.3 | NP_066978.1 | 7 | 1230 | |||
NR_026692.1 | 6 | 1330 | ||||
XM_005272472.4 | XP_005272529.1 | 7 | 847 | |||
GenBank mRNA | AF290886.1 | - | 7 | 1172 | ||
AK293089.1 | - | 7 | 1186 | |||
AK304190.1 | - | 7 | 733 | |||
AK308595.1 | - | 4 | 773 | |||
AK313585.1 | - | 7 | 1231 | |||
AY042221.1 | - | 7 | 1163 | |||
AY042222.1 | - | 7 | 1145 | |||
AY042223.1 | - | 7 | 887 | |||
AY042224.1 | - | 6 | 455 | |||
AY042225.1 | - | 6 | 1091 | |||
AY042226.1 | - | 5 | 1031 | |||
AY042227.1 | - | 6 | 755 | |||
AY042229.1 | - | 6 | 1263 | |||
AY042230.1 | - | 5 | 1191 | |||
AY042231.1 | - | 5 | 1053 | |||
BC110615.2 | - | 5 | 1198 | 40032459 | ||
M98457.1 | - | 7 | 1204 |