Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDR2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014603.2 | NP_055418.2 | 2 | 569 | ||
XM_006721852.2 | XP_006721915.1 | 2 | 554 | |||
GenBank mRNA | AK294006.1 | - | 2 | 569 | ||
BC047534.1 | - | 2 | 569 | 5288296 | ||
L02867.1 | - | 2 | 367 |