Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127207.1 | NP_001120679.1 | 6 | 1260 | ||
NM_014140.3 | NP_054859.2 | 6 | 1446 | |||
XM_005246631.2 | XP_005246688.1 | 6 | 1384 | |||
XM_005246632.1 | XP_005246689.1 | 6 | 1436 | |||
XM_017004228.1 | XP_016859717.1 | 6 | 1249 | |||
GenBank mRNA | AF082179.1 | - | 5 | 1216 | ||
AF432223.1 | - | 6 | 1256 | |||
AK023780.1 | - | 6 | 1387 | |||
AL122076.1 | - | 4 | 680 | |||
BC029994.1 | - | 4 | 708 | |||
BC043341.1 | - | 6 | 1233 | 6058575 |