Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127207.1 | NP_001120679.1 | 3 | 804 | ||
NM_014140.3 | NP_054859.2 | 3 | 990 | |||
XM_005246631.2 | XP_005246688.1 | 3 | 928 | |||
XM_005246632.1 | XP_005246689.1 | 3 | 980 | |||
XM_017004228.1 | XP_016859717.1 | 3 | 789 | |||
GenBank mRNA | AF082179.1 | - | 2 | 760 | ||
AF432223.1 | - | 3 | 800 | |||
AK023780.1 | - | 3 | 931 | |||
AL122076.1 | - | 1 | 224 | |||
BC016482.1 | - | 2 | 751 | 4341673 | ||
BC029994.1 | - | 1 | 252 | |||
BC043341.1 | - | 3 | 777 | 6058575 |