Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024937.3 | NP_001020108.1 | 32 | 4144 | ||
NM_001321236.1 | NP_001308165.1 | 32 | 4209 | |||
NM_015716.4 | NP_056531.1 | 31 | 4093 | |||
NM_153827.4 | NP_722549.2 | 32 | 4204 | |||
NM_170663.4 | NP_733763.1 | 32 | 4117 | |||
XM_006721531.2 | XP_006721594.1 | 33 | 4291 | |||
XM_006721532.2 | XP_006721595.1 | 33 | 4288 | |||
XM_006721536.2 | XP_006721599.1 | 32 | 4180 | |||
XM_011523906.1 | XP_011522208.1 | 32 | 4195 | |||
XM_017024704.1 | XP_016880193.1 | 33 | 4282 | |||
XM_017024705.1 | XP_016880194.1 | 33 | 4231 | |||
XM_017024706.1 | XP_016880195.1 | 32 | 4177 | |||
XM_017024707.1 | XP_016880196.1 | 33 | 4092 | |||
XM_017024708.1 | XP_016880197.1 | 31 | 3942 | |||
GenBank mRNA | AB035698.1 | - | 31 | 3849 | ||
AB041926.1 | - | 32 | 4057 | |||
AB209450.1 | - | 31 | 4742 | |||
AF218033.1 | - | 10 | 1222 | |||
AK096610.1 | - | 5 | 2338 | |||
AL157418.1 | - | 27 | 3756 | |||
AY775058.1 | - | 32 | 3900 | |||
BC034673.1 | - | 32 | 4156 | 4384442 |