Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015719.3 | NP_056534.2 | 13, 14 | 1506 | ||
XM_011528042.2 | XP_011526344.1 | 13, 14 | 1564 | |||
GenBank mRNA | AF177941.1 | - | 13, 14 | 1506 |