Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015719.3 | NP_056534.2 | 59 | 4305 | ||
XM_011528042.2 | XP_011526344.1 | 59 | 4363 | |||
XM_017026849.1 | XP_016882338.1 | 32 | 2006 | |||
GenBank mRNA | AB209175.1 | - | 57 | 3958 | ||
AF177941.1 | - | 59 | 4305 |