Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173982.1 | NP_001167453.1 | 3 | 1397 | ||
NM_018413.5 | NP_060883.1 | 3 | 1412 | |||
XM_017019369.1 | XP_016874858.1 | 5 | 983 | |||
GenBank mRNA | AB042326.1 | - | 3 | 1300 | ||
AF239820.1 | - | 3 | 1063 | |||
AJ269537.2 | - | 3 | 981 | |||
AJ289134.1 | - | 3 | 1039 | |||
AK290923.1 | - | 3 | 1353 | |||
AK313217.1 | - | 3 | 1340 | |||
BC013315.1 | - | 4 | 1209 | 4042813 |