Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018724.3 | NP_061194.2 | 3 | 386 | ||
XM_005273152.3 | XP_005273209.1 | 4 | 812 | |||
GenBank mRNA | AF224266.1 | - | 3 | 386 | ||
AY358320.1 | - | 4 | 705 | |||
AY876922.1 | - | 3 | 342 | |||
BC069311.1 | - | 3 | 342 | 7262116 | ||
BC069364.1 | - | 3 | 342 | 7262128 | ||
BC069398.1 | - | 3 | 342 | 7262140 | ||
BC069425.1 | - | 3 | 342 | 7262152 | ||
BC069449.1 | - | 3 | 342 | 7262164 | ||
BC069487.1 | - | 3 | 342 | 7262176 | ||
BC069523.1 | - | 3 | 342 | 7262093 | ||
BC069559.1 | - | 3 | 342 | 7262105 | ||
BC074948.2 | - | 3 | 367 | 30915498 | ||
BC074949.2 | - | 3 | 367 | 30915275 | ||
BC117386.1 | - | 3 | 368 | 40125937 | ||
BC117390.1 | - | 3 | 368 | 40125941 |