Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL21R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021798.3 | NP_068570.1 | 4 | 363 | ||
NM_181078.2 | NP_851564.1 | 4 | 716 | |||
NM_181079.4 | NP_851565.4 | 5 | 857 | |||
XM_011545857.2 | XP_011544159.1 | 5 | 893 | |||
XM_011545858.2 | XP_011544160.1 | 5 | 857 | |||
XM_017023257.1 | XP_016878746.1 | 5 | 846 | |||
GenBank mRNA | AF254067.1 | - | 4 | 311 | ||
AF269133.1 | - | 4 | 379 | |||
AK292663.1 | - | 4 | 716 | |||
AK312825.1 | - | 4 | 705 | |||
AY358826.1 | - | 4 | 346 | |||
BC004348.1 | - | 4 | 328 | 3634520 | ||
BQ215949.1 | - | 4 | 376 | |||
BX401767.2 | - | 4 | 476 | |||
DB115361.1 | - | 5 | 504 |