Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016287.3 | NP_057371.2 | 13 | 1572 | ||
XM_005245875.4 | XP_005245932.1 | 13 | 2147 | |||
XM_005245876.3 | XP_005245933.1 | 13 | 1680 | |||
XM_005245877.4 | XP_005245934.1 | 13 | 2008 | |||
XM_005245878.4 | XP_005245935.1 | 13 | 1944 | |||
XM_005245879.4 | XP_005245936.1 | 12 | 1471 | |||
XM_011541532.1 | XP_011539834.1 | 13 | 1664 | |||
XM_011541533.1 | XP_011539835.1 | 13 | 1861 | |||
XM_011541534.2 | XP_011539836.1 | 12 | 1481 | |||
XM_017001393.1 | XP_016856882.1 | 13 | 1627 | |||
XM_017001394.1 | XP_016856883.1 | 13 | 1992 | |||
GenBank mRNA | AF113534.1 | - | 12 | 1505 | ||
AK023129.1 | - | 13 | 1572 | |||
AK291252.1 | - | 13 | 1550 | |||
AK303456.1 | - | 6 | 688 | |||
AK304065.1 | - | 12 | 1448 | |||
AL833978.1 | - | 9 | 1388 | |||
BC045660.1 | - | 9 | 2204 | 5271429 | ||
BM312215.1 | - | 2 | 320 |