Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F11R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016946.4 | NP_058642.1 | 4 | 537 | ||
GenBank mRNA | AF154005.1 | - | 4 | 209 | ||
AF172398.2 | - | 4 | 413 | |||
AF207907.1 | - | 4 | 282 | |||
AK026665.1 | - | 4 | 328 | |||
AK075152.1 | - | 4 | 337 | |||
AK299487.1 | - | 4 | 318 | |||
AL136649.1 | - | 4 | 341 | |||
AY358896.1 | - | 4 | 318 | |||
BC001533.2 | - | 4 | 311 | 2990019 | ||
BT020103.1 | - | 4 | 267 | |||
CR533512.1 | - | 4 | 267 |