Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109878.1 | NP_001103348.1 | 5 | 716 | ||
NM_001109879.1 | NP_001103349.1 | 5 | 712 | |||
NM_001303475.1 | NP_001290404.1 | 3 | 470 | |||
NM_016954.2 | NP_058650.1 | 4 | 609 | |||
XM_005262136.2 | XP_005262193.1 | 5 | 744 | |||
XM_006724657.3 | XP_006724720.1 | 5 | 745 | |||
XM_011530972.1 | XP_011529274.1 | 4 | 1349 | |||
GenBank mRNA | AF251684.1 | - | 3 | 461 | ||
AK097514.1 | - | 5 | 712 | |||
AY035371.1 | - | 4 | 588 | |||
BC014194.2 | - | 4 | 609 | 3952809 |