Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016060.2 | NP_057144.1 | 3 | 294 | ||
GenBank mRNA | AF151883.1 | - | 3 | 266 | ||
AK094033.1 | - | 1 | 1577 | |||
AK311875.1 | - | 3 | 263 | |||
BC012539.1 | - | 3 | 269 | 4248123 |