Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016049.3 | NP_057133.2 | 5 | 562 | ||
XM_005267721.4 | XP_005267778.1 | 4 | 1530 | |||
GenBank mRNA | AF151870.1 | - | 5 | 557 | ||
AY368908.1 | - | 5 | 507 | |||
BC002491.2 | - | 5 | 562 | 3051105 |