Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016053.3 | NP_057137.1 | 4, 5 | 496 | ||
XM_011538423.1 | XP_011536725.1 | 4, 5 | 414 | |||
XM_017019383.1 | XP_016874872.1 | 3, 4 | 311 | |||
GenBank mRNA | AF151874.1 | - | 4, 5 | 441 | ||
AF155655.1 | - | 4, 5 | 441 | |||
AK223339.1 | - | 4, 5 | 432 | |||
AK314970.1 | - | 4, 5 | 414 | |||
CR457171.1 | - | 4, 5 | 334 |