Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016065.3 | NP_057149.1 | 2 | 428 | ||
XM_017016305.1 | XP_016871794.1 | 2 | 423 | |||
GenBank mRNA | AB049948.1 | - | 2 | 314 | ||
AF151890.1 | - | 2 | 381 | |||
AK025715.1 | - | 2 | 361 | |||
AK098291.1 | - | 2 | 374 | |||
BC021106.1 | - | 2 | 323 | 5020046 |