Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DESI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297746.1 | NP_001284675.1 | 4 | 738 | ||
NM_016076.4 | NP_057160.2 | 5 | 837 | |||
NR_123735.1 | 5 | 832 | ||||
NR_123736.1 | 4 | 743 | ||||
XM_011544203.2 | XP_011542505.1 | 6 | 922 | |||
XM_011544205.2 | XP_011542507.1 | 5 | 1488 | |||
XM_011544206.2 | XP_011542508.1 | 5 | 561 | |||
XM_011544207.2 | XP_011542509.1 | 4 | 1928 | |||
GenBank mRNA | AF151904.1 | - | 5 | 626 | ||
AF229834.1 | - | 4 | 479 | |||
AK314664.1 | - | 5 | 807 | |||
BC004485.2 | - | 5 | 784 | 3831754 | ||
CB854052.1 | - | 4 | 495 |