Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251989.1 | NP_001238918.1 | 7 | 821 | ||
NM_001251990.1 | NP_001238919.1 | 7 | 745 | |||
NM_001251991.1 | NP_001238920.1 | 7 | 729 | |||
NM_015895.4 | NP_056979.1 | 7 | 953 | |||
XM_005249159.1 | XP_005249216.1 | 7 | 992 | |||
XM_011514651.2 | XP_011512953.1 | 6 | 3047 | |||
GenBank mRNA | AF067855.1 | - | 6 | 639 | ||
AK021685.1 | - | 7 | 821 | |||
AK222992.1 | - | 6 | 612 | |||
AK315743.1 | - | 7 | 934 | |||
BC005185.1 | - | 7 | 876 | 4101771 | ||
BC005389.1 | - | 7 | 928 | 3996265 | ||
CB131959.1 | - | 4 | 443 |