Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 15 | 3643 | ||
XM_011517056.2 | XP_011515358.1 | 16 | 4461 | |||
XM_011517058.2 | XP_011515360.1 | 15 | 4316 | |||
XM_011517059.1 | XP_011515361.1 | 15 | 3583 | |||
XM_011517060.2 | XP_011515362.1 | 16 | 4443 | |||
XM_011517061.2 | XP_011515363.1 | 16 | 3800 | |||
XM_011517062.1 | XP_011515364.1 | 15 | 3655 | |||
XM_011517063.2 | XP_011515365.1 | 15 | 4298 | |||
XM_011517064.2 | XP_011515366.1 | 16 | 4448 | |||
XM_011517065.1 | XP_011515367.1 | 15 | 3719 | |||
XM_011517068.2 | XP_011515370.1 | 15 | 3536 | |||
XM_011517069.2 | XP_011515371.1 | 10 | 3642 | |||
XM_011517070.2 | XP_011515372.1 | 16 | 4465 | |||
XM_011517072.2 | XP_011515374.1 | 9 | 2900 | |||
XM_011517073.2 | XP_011515375.1 | 9 | 2904 | |||
XM_011517074.1 | XP_011515376.1 | 8 | 2770 | |||
XM_017013470.1 | XP_016868959.1 | 15 | 4255 | |||
XM_017013471.1 | XP_016868960.1 | 16 | 4404 | |||
GenBank mRNA | AB196635.1 | - | 15 | 3540 | ||
AF131744.1 | - | 3 | 1298 | |||
BC037288.1 | - | 16 | 3678 | 5266568 | ||
BC106912.1 | - | 3 | 1209 | |||
BC106913.1 | - | 3 | 1209 | |||
CR457147.1 | - | 3 | 968 | |||
CR936831.1 | - | 9 | 2836 |