Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015913.3 | NP_056997.1 | 4 | 1305 | ||
GenBank mRNA | AF131758.1 | - | 4 | 340 | ||
AF543416.1 | - | 4 | 509 | |||
AK075409.1 | - | 4 | 303 | |||
AY358982.1 | - | 4 | 336 | |||
BC001493.2 | - | 4 | 306 | 3506109 | ||
BC008913.2 | - | 4 | 335 | 3659407 | ||
BC008953.2 | - | 4 | 284 | 2959502 |