Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127713.1 | NP_001121185.1 | 3 | 562 | ||
NM_015915.4 | NP_056999.2 | 2 | 378 | |||
NM_181598.3 | NP_853629.2 | 2 | 378 | |||
GenBank mRNA | AF444143.1 | - | 2 | 309 | ||
AK223436.1 | - | 2 | 291 | |||
AK290185.1 | - | 2 | 291 | |||
AK312518.1 | - | 2 | 267 | |||
AL833591.1 | - | 3 | 346 | |||
AY032844.1 | - | 2 | 306 | |||
BG704589.1 | - | 2 | 366 | |||
BI601372.1 | - | 2 | 381 | |||
DA088230.1 | - | 2 | 378 | |||
KU178621.1 | - | 2 | 137 | |||
KU178622.1 | - | 2 | 137 | |||
KU178623.1 | - | 2 | 137 | |||
KU178624.1 | - | 2 | 137 |