Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127713.1 | NP_001121185.1 | 3, 4 | 701 | ||
NM_015915.4 | NP_056999.2 | 2, 3 | 517 | |||
NM_181598.3 | NP_853629.2 | 2, 3 | 517 | |||
GenBank mRNA | AF131801.1 | - | 1, 2 | 41 | ||
AF444143.1 | - | 2, 3 | 448 | |||
AK223436.1 | - | 2, 3 | 430 | |||
AK290185.1 | - | 2, 3 | 430 | |||
AK312518.1 | - | 2, 3 | 406 | |||
AL833591.1 | - | 3, 4 | 485 | |||
AY032844.1 | - | 2, 3 | 445 | |||
BC010708.2 | - | 2, 3 | 420 | 3869877 | ||
BG704589.1 | - | 2, 3 | 507 | |||
BI601372.1 | - | 2, 3 | 522 | |||
CR457153.1 | - | 1, 2 | 27 | |||
DA088230.1 | - | 2, 3 | 517 | |||
KU178621.1 | - | 2, 3 | 276 | |||
KU178622.1 | - | 2, 3 | 276 | |||
KU178623.1 | - | 2, 3 | 276 |