Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320227.1 | NP_001307156.1 | 12 | 1192 | ||
NM_015938.4 | NP_057022.2 | 12 | 1192 | |||
XM_005247511.1 | XP_005247568.1 | 12 | 1142 | |||
XM_005247512.1 | XP_005247569.1 | 12 | 1473 | |||
GenBank mRNA | AF132941.1 | - | 12 | 1163 | ||
AK021775.1 | - | 9 | 803 | |||
AK094749.1 | - | 11 | 1495 | |||
AK225664.1 | - | 12 | 1174 | |||
AK296710.1 | - | 11 | 800 | |||
AK308865.1 | - | 11 | 973 | |||
BC013317.1 | - | 12 | 1175 | 4291797 |