Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300848.1 | NP_001287777.1 | 3 | 516 | ||
NM_015950.4 | NP_057034.2 | 3 | 516 | |||
XM_005249161.4 | XP_005249218.1 | 3 | 337 | |||
GenBank mRNA | AF132956.1 | - | 3 | 390 | ||
AK301041.1 | - | 3 | 373 | |||
AK314948.1 | - | 3 | 374 | |||
BC013685.1 | - | 3 | 361 | 3862065 | ||
BC020212.1 | - | 3 | 361 | 4641018 | ||
BM563754.1 | - | 482 | ||||
BM970997.1 | - | 4 | 548 |