Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015993.2 | NP_057077.1 | 3, 4 | 573 | ||
GenBank mRNA | AF137386.1 | - | 3, 4 | 569 | ||
AK130584.1 | - | 3, 4 | 565 | |||
AK289599.1 | - | 3, 4 | 550 | |||
BC002760.2 | - | 3, 4 | 536 | 3633345 |