Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077180.2 | NP_001070648.1 | 5 | 1125 | ||
NM_001288659.1 | NP_001275588.1 | 5 | 785 | |||
NM_001288660.1 | NP_001275589.1 | 5 | 782 | |||
NM_016025.4 | NP_057109.3 | 5 | 1128 | |||
XM_017023269.1 | XP_016878758.1 | 4 | 821 | |||
GenBank mRNA | AF151839.1 | - | 5 | 789 | ||
AJ278577.1 | - | 5 | 1046 | |||
AJ278578.1 | - | 5 | 1043 | |||
AK025565.1 | - | 3 | 359 | |||
AK074529.1 | - | 5 | 1024 | |||
AK075022.1 | - | 5 | 1011 | |||
AK075237.1 | - | 5 | 994 | |||
AK313077.1 | - | 5 | 1012 | |||
BC000195.1 | - | 5 | 1013 | 3350598 | ||
BX647201.1 | - | 1 | 3475 |