Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016037.3 | NP_057121.2 | 6 | 642 | ||
XM_011541555.2 | XP_011539857.1 | 5 | 576 | |||
GenBank mRNA | AF067802.1 | - | 6 | 605 | ||
AF151852.1 | - | 6 | 620 | |||
AK023587.1 | - | 4 | 1511 | |||
AK291900.1 | - | 6 | 572 | |||
AK308377.1 | - | 5 | 577 | |||
BC005182.1 | - | 6 | 587 | 3995984 |