Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016094.3 | NP_057178.2 | 1 | 74 | ||
NR_135149.1 | 1 | 74 | ||||
GenBank mRNA | AF059618.1 | - | 1 | 50 | ||
AF125096.1 | - | 1 | 60 | |||
AY542158.1 | - | 1 | 19 | |||
BC001228.1 | - | 1 | 53 | |||
BC022206.1 | - | 1 | 58 | |||
BC046131.1 | - | 1 | 36 | 4443942 | ||
BC093077.1 | - | 1 | 64 | 30372667 | ||
BG700015.1 | - | 1 | 77 | |||
BI792102.1 | - | 1 | 82 | |||
CN483677.1 | - | 1 | 25 | |||
DA811158.1 | - | 1 | 73 |