Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016100.4 | NP_057184.1 | 4 | 369 | ||
NM_181527.3 | NP_852668.1 | 4 | 456 | |||
NM_181528.3 | NP_852669.1 | 4 | 369 | |||
GenBank mRNA | AF085355.1 | - | 4 | 332 | ||
AK311819.1 | - | 4 | 322 | |||
AL136641.1 | - | 4 | 301 | |||
BC005181.1 | - | 4 | 297 | 3950192 | ||
BC008446.1 | - | 4 | 318 | 4104421 | ||
BG548527.1 | - | 4 | 317 | |||
BX338359.1 | - | 3 | 653 | |||
CB959658.1 | - | 4 | 393 |