Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETDB2-PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320727.1 | NP_001307656.1 | 17, 18 | 3032 | ||
NR_135324.1 | 17, 18 | 3071 |
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040443.2 | NP_001035533.1 | 7, 8 | 1036 | ||
NM_001040444.2 | NP_001035534.1 | 7, 8 | 712 | |||
NR_135322.1 | 6, 7 | 914 | ||||
NR_135323.1 | 8, 9 | 848 | ||||
GenBank mRNA | AB011031.1 | - | 7, 8 | 685 | ||
AF155105.1 | - | 7, 8 | 655 | |||
AK026228.1 | - | 7, 8 | 676 | |||
AK225986.1 | - | 8, 9 | 798 | |||
BC017212.2 | - | 7, 8 | 665 | 4054134 | ||
BX461254.2 | - | 6, 7 | 544 |