Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016120.3 | NP_057204.2 | 2 | 316 | ||
NM_183353.2 | NP_899196.1 | 3 | 378 | |||
GenBank mRNA | AF155109.1 | - | 2 | 266 | ||
AK001334.1 | - | 2 | 298 | |||
AL712605.1 | - | 3 | 360 | |||
BC013357.2 | - | 2 | 280 | 3834870 | ||
BF686000.1 | - | 2 | 316 |