Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRAK4 SIRNA Assays ›
Gene Symbol:
IRAK4
Gene Name:
interleukin 1 receptor associated kinase 4
Gene Aliases:
IPD1, IRAK-4, NY-REN-64, REN64
Chromosome Location:
Chr.12: 43758913 - 43789543 on Build GRCh38
UniGene ID:
Hs.138499
Species:
Human
siRNA ID
HSS121734
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRAK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114182.2
NP_001107654.1
8
942
NM_001145256.1
NP_001138728.1
7
796
NM_001145257.1
NP_001138729.1
6
748
NM_001145258.1
NP_001138730.1
5
578
NM_016123.3
NP_057207.2
7
894
XM_005268943.3
XP_005269000.1
7
999
XM_005268944.4
XP_005269001.1
7
963
XM_005268945.4
XP_005269002.1
7
1207
XM_005268947.4
XP_005269004.1
8
953
XM_005268948.2
XP_005269005.1
9
993
XM_005268949.2
XP_005269006.1
6
853
XM_006719438.3
XP_006719501.1
8
1255
XM_006719439.2
XP_006719502.1
7
1360
XM_006719440.2
XP_006719503.1
7
771
XM_006719441.2
XP_006719504.1
6
656
XM_006719442.2
XP_006719505.1
7
660
XM_006719443.3
XP_006719506.1
6
803
XM_011538431.2
XP_011536733.1
8
1011
XM_011538432.1
XP_011536734.1
8
1047
XM_011538433.2
XP_011536735.1
7
1626
XM_017019390.1
XP_016874879.1
9
1128
XM_017019391.1
XP_016874880.1
8
1093
GenBank mRNA
AF155118.1
-
7
861
AF445802.1
-
6
812
AK000528.1
-
7
841
AK027301.2
-
7
1389
AK299944.1
-
6
745
AK304583.1
-
5
581
AK314288.1
-
7
883
AY283670.1
-
6
812
AY283671.1
-
6
810
AY340962.1
-
7
877
AY340963.1
-
8
925
AY340964.1
-
6
731
AY340965.1
-
7
779
AY340966.1
-
8
944
AY340967.1
-
9
992
AY730682.1
-
6
812
BC013316.1
-
7
882
4287014
KU178631.1
-
6
812
KU178632.1
-
5
666
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
toll-like receptor signaling pathway
neutrophil mediated immunity
MyD88-dependent toll-like receptor signaling pathway
JNK cascade
cytokine-mediated signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of smooth muscle cell proliferation
neutrophil migration
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-nvdhv:80/10.239.163.173:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0