Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRTO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016183.3 | NP_057267.2 | 8 | 856 | ||
XM_006710675.3 | XP_006710738.1 | 8 | 1014 | |||
GenBank mRNA | AF173378.1 | - | 8 | 631 | ||
AK024227.1 | - | 8 | 617 | |||
AK295328.1 | - | 7 | 911 | |||
AY303790.1 | - | 8 | 612 | |||
BC003013.1 | - | 8 | 630 | 2821148 | ||
BC006504.2 | - | 8 | 630 | 2821148 |