Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135643.1 | NP_001129115.1 | 9, 10 | 958 | ||
NM_001135644.1 | NP_001129116.1 | 8, 9 | 1105 | |||
NM_016221.3 | NP_057305.1 | 8, 9 | 937 | |||
XM_005268457.1 | XP_005268514.1 | 7, 8 | 831 | |||
XM_011537644.1 | XP_011535946.1 | 9, 10 | 841 | |||
XM_011537645.1 | XP_011535947.1 | 9, 10 | 753 | |||
XM_011537646.1 | XP_011535948.1 | 9, 10 | 974 | |||
XM_011537647.1 | XP_011535949.1 | 9, 10 | 797 | |||
GenBank mRNA | AF195120.1 | - | 8, 9 | 852 | ||
AK000299.1 | - | 8, 9 | 856 | |||
AK125973.1 | - | 8, 9 | 1105 | |||
AK299517.1 | - | 8, 9 | 858 | |||
AK307757.1 | - | 9, 10 | 897 | |||
BC026323.1 | - | 8, 9 | 862 | 4815273 | ||
KU178637.1 | - | 8, 9 | 835 | |||
KU178638.1 | - | 8, 9 | 835 | |||
KU178639.1 | - | 5, 6 | 646 |