Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016299.3 | NP_057383.2 | 10 | 1220 | ||
GenBank mRNA | AF112210.1 | - | 10 | 1039 | ||
AF143723.1 | - | 10 | 1029 | |||
AK225265.1 | - | 10 | 1064 | |||
AK225651.1 | - | 10 | 1064 | |||
AK226065.1 | - | 3 | 3893 | |||
AK292323.1 | - | 10 | 1028 | |||
BC119690.1 | - | 10 | 1039 | 40116697 |