Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173482.1 | NP_001166953.1 | 2 | 133 | ||
NM_016302.3 | NP_057386.2 | 2 | 136 | |||
XM_011533791.2 | XP_011532093.1 | 2 | 123 | |||
XM_011533793.2 | XP_011532095.1 | 2 | 123 | |||
GenBank mRNA | AF117230.1 | - | 2 | 100 | ||
AK027507.1 | - | 2 | 114 | |||
AK058191.1 | - | 2 | 110 | |||
AK312577.1 | - | 2 | 114 | |||
AL711072.1 | - | 2 | 126 | |||
BC017419.1 | - | 2 | 124 | 4672890 | ||
BC067811.1 | - | 2 | 117 | 5264951 | ||
DB456585.1 | - | 2 | 136 |