Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL24D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016304.2 | NP_057388.1 | 2 | 319 | ||
GenBank mRNA | AB462502.1 | - | 2 | 203 | ||
AF060926.1 | - | 2 | 186 | |||
AF165521.1 | - | 2 | 319 | |||
AF201949.1 | - | 2 | 319 | |||
AF212226.1 | - | 2 | 319 | |||
AK055713.1 | - | 2 | 206 | |||
AK315431.1 | - | 2 | 219 | |||
BC008409.1 | - | 2 | 241 | 4097130 | ||
BC008422.1 | - | 2 | 211 | 4096889 | ||
BC008449.1 | - | 2 | 217 | 4133589 | ||
BC008499.1 | - | 2 | 232 | 4286115 | ||
BC009593.1 | - | 2 | 222 | 4109097 | ||
BC009604.1 | - | 2 | 222 | 4282059 | ||
BC012913.1 | - | 2 | 186 | 4514979 | ||
BC014576.1 | - | 2 | 221 | 3930303 | ||
BC016312.1 | - | 2 | 227 | 4096867 | ||
BC016331.1 | - | 2 | 227 | 4108672 | ||
BC016725.1 | - | 2 | 228 | 4078305 | ||
BC016777.1 | - | 2 | 209 | 4104810 | ||
BC026266.1 | - | 2 | 223 | 4276092 | ||
BC026267.1 | - | 2 | 202 | 4277062 | ||
BC028672.1 | - | 2 | 208 | 4092804 | ||
BC035995.1 | - | 2 | 232 | 4671856 | ||
BC093067.1 | - | 2 | 206 | 30419021 | ||
CR457179.1 | - | 2 | 176 |