Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020921.3 | NP_065972.3 | 18 | 3498 | ||
NM_182944.2 | NP_891989.2 | 18 | 3498 | |||
NM_182946.1 | NP_891991.1 | 18 | 3498 | |||
XM_011536817.1 | XP_011535119.1 | 18 | 3472 | |||
XM_011536819.2 | XP_011535121.1 | 18 | 3472 | |||
XM_011536823.2 | XP_011535125.1 | 17 | 3349 | |||
GenBank mRNA | AF212162.2 | - | 17 | 3527 | ||
AF223937.1 | - | 19 | 3457 | |||
AF223938.1 | - | 19 | 3457 | |||
AF223939.1 | - | 19 | 3457 |