Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016350.4 | NP_057434.4 | 21 | 3036 | ||
NM_020921.3 | NP_065972.3 | 22 | 5175 | |||
NM_182944.2 | NP_891989.2 | 22 | 5175 | |||
NM_182946.1 | NP_891991.1 | 22 | 5175 | |||
XM_011536817.1 | XP_011535119.1 | 22 | 5149 | |||
XM_011536819.2 | XP_011535121.1 | 22 | 5149 | |||
XM_011536822.1 | XP_011535124.1 | 21 | 3010 | |||
XM_011536823.2 | XP_011535125.1 | 21 | 5026 | |||
GenBank mRNA | AB046785.2 | - | 21 | 2958 | ||
AF186776.1 | - | 3 | 304 | |||
AF212162.2 | - | 21 | 5204 | |||
AF223937.1 | - | 23 | 5134 | |||
AF223938.1 | - | 23 | 5134 | |||
AF223939.1 | - | 23 | 5134 | |||
AF302773.2 | - | 22 | 5175 | |||
AK025652.1 | - | 3 | 295 | |||
AK026083.1 | - | 5 | 734 | |||
AY027794.1 | - | 3 | 304 | |||
AY027795.1 | - | 3 | 304 | |||
AY027796.1 | - | 3 | 304 | |||
AY736182.1 | - | 22 | 5175 |