Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163446.1 | NP_001156918.1 | 7 | 687 | ||
NM_016352.3 | NP_057436.2 | 8 | 786 | |||
GenBank mRNA | AF095719.1 | - | 8 | 746 | ||
AK290993.1 | - | 8 | 786 | |||
AK298550.1 | - | 7 | 658 | |||
AK298553.1 | - | 6 | 472 | |||
AK299023.1 | - | 6 | 540 | |||
AY358699.1 | - | 8 | 747 | |||
BC052289.1 | - | 8 | 767 | 6106874 |