Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016355.3 | NP_057439.2 | 10, 11 | 1132 | ||
NM_201224.1 | NP_957518.1 | 9, 10 | 985 | |||
GenBank mRNA | AF078843.1 | - | 9, 10 | 942 | ||
AF190165.1 | - | 9, 10 | 1645 | |||
AJ276704.1 | - | 10, 11 | 1110 | |||
AK054574.1 | - | 10, 11 | 1117 | |||
AK222940.1 | - | 10, 11 | 1091 | |||
AK225144.1 | - | 10, 11 | 1102 | |||
AL136666.1 | - | 10, 11 | 1136 | |||
AL833469.1 | - | 7, 8 | 2005 | |||
BC009379.2 | - | 10, 11 | 1095 | |||
BC068009.1 | - | 10, 11 | 1114 | 6142837 |