Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016355.3 | NP_057439.2 | 5 | 545 | ||
NM_201224.1 | NP_957518.1 | 5 | 545 | |||
GenBank mRNA | AF078843.1 | - | 5 | 502 | ||
AF190165.1 | - | 5 | 509 | |||
AJ276704.1 | - | 5 | 523 | |||
AK054574.1 | - | 5 | 530 | |||
AK222940.1 | - | 5 | 504 | |||
AK225144.1 | - | 5 | 515 | |||
AK310501.1 | - | 5 | 575 | |||
AL136666.1 | - | 5 | 549 | |||
AL833469.1 | - | 4 | 345 | |||
BC009379.2 | - | 5 | 508 | |||
BC068009.1 | - | 5 | 527 | 6142837 | ||
EF036507.1 | - | 5 | 492 |