Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323625.1 | NP_001310554.1 | 9 | 2447 | ||
NM_016361.4 | NP_057445.4 | 10 | 1737 | |||
NR_136633.1 | 9 | 2402 | ||||
NR_136634.1 | 10 | 2777 | ||||
NR_136635.1 | 10 | 1723 | ||||
NR_136636.1 | 9 | 2722 | ||||
GenBank mRNA | AB030039.2 | - | 10 | 1541 | ||
AB031478.1 | - | 10 | 1648 | |||
AB209248.1 | - | 8 | 4498 | |||
AK000657.1 | - | 10 | 1474 | |||
AY358552.1 | - | 10 | 1445 | |||
BC009965.1 | - | 10 | 1717 | 4299943 | ||
BC034686.1 | - | 10 | 1721 | 3463770 |